- Pyruvate kinase type M2 and its role in tumor growth and spreading.
Pyruvate kinase type M2 and its role in tumor growth and spreading.
Seminars in cancer biology (2005-05-24)
Sybille Mazurek, C Bruce Boschek, Ferdinand Hugo, Erich Eigenbrodt
PMID15908230
ABSTRACT
Proliferating cells and tumor cells in particular express the pyruvate kinase isoenzyme type M2 (M2-PK). Within the tumor metabolome M2-PK regulates the proportions of glucose carbons that are channelled to synthetic processes (inactive dimeric form) or used for glycolytic energy production (highly active tetrameric form, a component of the glycolytic enzyme complex). In tumor cells, the dimeric form of M2-PK (Tumor M2-PK) is always predominant. The dimerization is caused by direct interaction of M2-PK with certain oncoproteins. The switch between the tetrameric and dimeric form of M2-PK allows tumor cells to survive in environments with varying oxygen und nutrient supply.