Merck
  • Antibodies bound to Aβ oligomers potentiate the neurotoxicity of Aβ by activating microglia.
Antibodies bound to Aβ oligomers potentiate the neurotoxicity of Aβ by activating microglia.
Journal of neurochemistry (2013-06-12)
Ramune Morkuniene, Aurelija Zvirbliene, Indre Dalgediene, Paulius Cizas, Silvija Jankeviciute, Giedre Baliutyte, Ramunas Jokubka, Marija Jankunec, Gintaras Valincius, Vilmante Borutaite
ABSTRACT
Beta amyloid (Aβ) oligomers are thought to contribute to the pathogenesis of Alzheimer’s disease. However, clinical trials using Aβ immunization were unsuccessful due to strong brain inflammation, the mechanisms of which are poorly understood. In this study we tested whether monoclonal antibodies to oligomeric Aβ would prevent the neurotoxicity of Aβ oligomers in primary neuronal-glial cultures. However, surprisingly,the antibodies dramatically increased the neurotoxicity of Aβ. Antibodies bound to monomeric Aβ fragments were non-toxic to cultured neurons, while antibodies to other oligomeric proteins: hamster polyomavirus major capsid protein, human metapneumovirus nucleocapsid protein, and measles virus nucleocapsid protein, strongly potentiated the neurotoxicity of their antigens. The neurotoxicity of antibody-antibody oligomeric antigen complexes was abolished by removal of the Fc region from the antibodies or by removal of microglia from cultures, and was accompanied by inflammatory activation and proliferation of the microglia in culture. In conclusion, we find that immune complexes formed by Aβ oligomers or other oligomeric/multimeric antigens and their specific antibodies can cause death and loss of neurons in primary neuronal-glial cultures via Fc-dependent microglial activation. The results suggest that therapies resulting in antibodies to oligomeric Aβ or oligomeric brain virus proteins should be used with caution or with suppression of microglial activation.
Sigma-Aldrich
Anti-Amyloid β42 Antibody, clone G2-13, clone G2-13, from mouse