Merck
  • Human Bocavirus 1 Infection of Well-Differentiated Human Airway Epithelium.
Human Bocavirus 1 Infection of Well-Differentiated Human Airway Epithelium.
Current protocols in microbiology (2020-07-09)
Ziying Yan, Xuefeng Deng, Jianming Qiu
ABSTRACT
Human bocavirus 1 (HBoV1) is a small DNA virus that belongs to the Bocaparvovirus genus of the Parvoviridae family. HBoV1 is a common respiratory pathogen that causes mild to life-threatening acute respiratory tract infections in children and immunocompromised individuals, infecting both the upper and lower respiratory tracts. HBoV1 infection causes death of airway epithelial cells, resulting in airway injury and inflammation. In vitro, HBoV1 only infects well-differentiated (polarized) human airway epithelium cultured at an air-liquid interface (HAE-ALI), but not any dividing human cells. A full-length HBoV1 genome of 5543 nucleotides has been cloned from DNA extracted from a human nasopharyngeal swab into a plasmid called HBoV1 infectious clone pIHBoV1. Transfection of pIHBoV1 replicates efficiently in human embryonic kidney 293 (HEK293) cells and produces virions that are highly infectious. This article describes protocols for production of HBoV1 in HEK293 cells, generation of HAE-ALI cultures, and infection with HBoV1 in HAE-ALI. © 2020 Wiley Periodicals LLC. Basic Protocol 1: Human bocavirus 1 production in HEK293 cells Support Protocol 1: HEK293 cell culture and transfection Support Protocol 2: Quantification of human bocavirus 1 using real-time quantitative PCR Basic Protocol 2: Differentiation of human airway cells at an air-liquid interface Support Protocol 3: Expansion of human airway epithelial cell line CuFi-8 Support Protocol 4: Expansion of human airway basal cells Support Protocol 5: Coating of plastic dishes and permeable membranes of inserts Support Protocol 6: Transepithelial electrical resistance measurement Basic Protocol 3: Human bocavirus 1 infection in human airway epithelium cultured at an air-liquid interface Support Protocol 7: Isolation of infected human airway epithelium cells from inserts Basic Protocol 4: Transduction of airway basal cells with lentiviral vector.
Sigma-Aldrich
Gentamicin solution, 50 mg/mL in deionized water, liquid, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USDA approved, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Millipore
Benzonase® Nuclease, ≥250 units/μL, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution
Sigma-Aldrich
Amphotericin B solubilized, powder, γ-irradiated, BioXtra, suitable for cell culture