Merck
Not so sweet and simple: impacts of SARS-CoV-2 on the β cell.
Islets (2021-05-11)
Sarah Ibrahim, Gabriela S F Monaco, Emily K Sims
ABSTRACT
The link between COVID-19 infection and diabetes has been explored in several studies since the start of the pandemic, with associations between comorbid diabetes and poorer prognosis in patients infected with the virus and reports of diabetic ketoacidosis occurring with COVID-19 infection. As such, significant interest has been generated surrounding mechanisms by which the virus may exert effects on the pancreatic β cells. In this review, we consider possible routes by which SARS-CoV-2 may impact β cells. Specifically, we outline data that either support or argue against the idea of direct infection and injury of β cells by SARS-CoV-2. We also discuss β cell damage due to a "bystander" effect in which infection with the virus leads to damage to surrounding tissues that are essential for β cell survival and function, such as the pancreatic microvasculature and exocrine tissue. Studies elucidating the provocation of a cytokine storm following COVID-19 infection and potential impacts of systemic inflammation and increases in insulin resistance on β cells are also reviewed. Finally, we summarize the existing clinical data surrounding diabetes incidence since the start of the COVID-19 pandemic.
Sigma-Aldrich
Anti-ACE2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-TMPRSS2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution