Merck
  • Self-propelled nanomotor reconstructs tumor microenvironment through synergistic hypoxia alleviation and glycolysis inhibition for promoted anti-metastasis.
Self-propelled nanomotor reconstructs tumor microenvironment through synergistic hypoxia alleviation and glycolysis inhibition for promoted anti-metastasis.
Acta pharmaceutica Sinica. B (2021-10-01)
Wenqi Yu, Ruyi Lin, Xueqin He, Xiaotong Yang, Huilin Zhang, Chuan Hu, Rui Liu, Yuan Huang, Yi Qin, Huile Gao
ABSTRACT
Solid tumors always exhibit local hypoxia, resulting in the high metastasis and inertness to chemotherapy. Reconstruction of hypoxic tumor microenvironment (TME) is considered a potential therapy compared to directly killing tumor cells. However, the insufficient oxygen delivery to deep tumor and the confronting "Warburg effect" compromise the efficacy of hypoxia alleviation. Herein, we construct a cascade enzyme-powered nanomotor (NM-si), which can simultaneously provide sufficient oxygen in deep tumor and inhibit the aerobic glycolysis to potentiate anti-metastasis in chemotherapy. Catalase (Cat) and glucose oxidase (GOx) are co-adsorbed on our previously reported CAuNCs@HA to form self-propelled nanomotor (NM), with hexokinase-2 (HK-2) siRNA further condensed (NM-si). The persistent production of oxygen bubbles from the cascade enzymatic reaction propels NM-si to move forward autonomously and in a controllable direction along H2O2 gradient towards deep tumor, with hypoxia successfully alleviated in the meantime. The autonomous movement also facilitates NM-si with lysosome escaping for efficient HK-2 knockdown to inhibit glycolysis. In vivo results demonstrated a promising anti-metastasis effect of commercially available albumin-bound paclitaxel (PTX@HSA) after pre-treated with NM-si for TME reconstruction. This cascade enzyme-powered nanomotor provides a potential prospect in reversing the hypoxic TME and metabolic pathway for reinforced anti-metastasis of chemotherapy.
Sigma-Aldrich
Catalase from bovine liver, powder, suitable for cell culture, 2,000-5,000 units/mg protein