Merck
  • Spine impairment in mice high-expressing neuregulin 1 due to LIMK1 activation.
Spine impairment in mice high-expressing neuregulin 1 due to LIMK1 activation.
Cell death & disease (2021-04-16)
Peng Chen, Hongyang Jing, Mingtao Xiong, Qian Zhang, Dong Lin, Dongyan Ren, Shunqi Wang, Dongmin Yin, Yongjun Chen, Tian Zhou, Baoming Li, Erkang Fei, Bing-Xing Pan
ABSTRACT
The genes encoding for neuregulin1 (NRG1), a growth factor, and its receptor ErbB4 are both risk factors of major depression disorder and schizophrenia (SZ). They have been implicated in neural development and synaptic plasticity. However, exactly how NRG1 variations lead to SZ remains unclear. Indeed, NRG1 levels are increased in postmortem brain tissues of patients with brain disorders. Here, we studied the effects of high-level NRG1 on dendritic spine development and function. We showed that spine density in the prefrontal cortex and hippocampus was reduced in mice (ctoNrg1) that overexpressed NRG1 in neurons. The frequency of miniature excitatory postsynaptic currents (mEPSCs) was reduced in both brain regions of ctoNrg1 mice. High expression of NRG1 activated LIMK1 and increased cofilin phosphorylation in postsynaptic densities. Spine reduction was attenuated by inhibiting LIMK1 or blocking the NRG1-LIMK1 interaction, or by restoring NRG1 protein level. These results indicate that a normal NRG1 protein level is necessary for spine homeostasis and suggest a pathophysiological mechanism of abnormal spines in relevant brain disorders.
Sigma-Aldrich
Polyethylenimine, branched, average Mw ~25,000 by LS, average Mn ~10,000 by GPC, branched
Sigma-Aldrich
Anti-PSD95 Antibody, clone 3H4.3, clone 3H4.3, from mouse
Sigma-Aldrich
Pararosaniline Solution