Merck
  • Effects of TGF-β1 Receptor Inhibitor GW788388 on the Epithelial to Mesenchymal Transition of Peritoneal Mesothelial Cells.
Effects of TGF-β1 Receptor Inhibitor GW788388 on the Epithelial to Mesenchymal Transition of Peritoneal Mesothelial Cells.
International journal of molecular sciences (2021-05-06)
Yunmee Lho, Jun-Young Do, Jung-Yoon Heo, A-Young Kim, Sang-Woon Kim, Seok-Hui Kang
ABSTRACT
We investigated the effectiveness of the transforming growth factor beta-1 (TGF-β) receptor inhibitor GW788388 on the epithelial to mesenchymal transition (EMT) using human peritoneal mesothelial cells (HPMCs) and examined the effectiveness of GW788388 on the peritoneal membrane using a peritoneal fibrosis mouse model. HPMCs were treated with TGF-β with or without GW788388. Animal experiments were conducted on male C57/BL6 mice. Peritoneal fibrosis was induced by intraperitoneal injection of chlorhexidine gluconate. GW788388 was administered by once-daily oral gavage. The morphological change, cell migration, and invasion resulted from TGF-β treatment, but these changes were attenuated by cotreatment with GW788388. TGF-β-treated HPMCs decreased the level of the epithelial cell marker and increased the levels of the mesenchymal cell markers. Cotreatment with GW788388 reversed these changes. Phosphorylated Smad2 and Smad3 protein levels were stimulated with TGF-β and the change was attenuated by cotreatment with GW788388. For the peritoneal fibrosis mice, thickness and collagen deposition of parietal peritoneum was increased, but this change was attenuated by cotreatment with GW788388. GW788388, an orally available potent TGF-β receptor type 1 inhibitor, effectively attenuated TGF-β-induced EMT in HPMCs. Cotreatment with GW788388 improved peritoneal thickness and fibrosis, and recovered peritoneal membrane function in a peritoneal fibrosis mouse model.
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Crystal Violet Solution, 1%, aqueous solution
Sigma-Aldrich
GW788388 Hydrate, ≥98% (HPLC)