Merck
  • Adenosine deaminase 2 produced by infiltrative monocytes promotes liver fibrosis in nonalcoholic fatty liver disease.
Adenosine deaminase 2 produced by infiltrative monocytes promotes liver fibrosis in nonalcoholic fatty liver disease.
Cell reports (2021-10-28)
Shilpa Tiwari-Heckler, Eric U Yee, Yusuf Yalcin, Jiwoon Park, Duc-Huy T Nguyen, Wenda Gao, Eva Csizmadia, Nezam Afdhal, Kenneth J Mukamal, Simon C Robson, Michelle Lai, Robert E Schwartz, Z Gordon Jiang
ABSTRACT
Elevated circulating activity of adenosine deaminase 2 (ADA2) is associated with liver fibrosis in nonalcoholic fatty liver disease (NAFLD). In the liver of NAFLD patients, ADA2-positive portal macrophages are significantly associated with the degree of liver fibrosis. These liver macrophages are CD14- and CD16-positive and co-express chemokine receptors CCR2, CCR5, and CXCR3, indicating infiltrative monocyte origin. Human circulatory monocytes release ADA2 upon macrophage differentiation in vitro. When stimulated by recombinant human ADA2 (rhADA2), human monocyte-derived macrophages demonstrate upregulation of pro-inflammatory and pro-fibrotic genes, including PDGF-B, a key pro-fibrotic cytokine. This PDGF-B upregulation is reproduced by inosine, the enzymatic product of ADA2, but not adenosine, and is abolished by E359N, a loss-of-function mutation in ADA2. Finally, rhADA2 also stimulates PDGF-B production from Kupffer cells in primary human liver spheroids. Together, these data suggest that infiltrative monocytes promote fibrogenesis in NAFLD via ADA2-mediated autocrine/paracrine signaling culminating in enhanced PDGF-B production.