Merck
  • Snapshots of the Signaling Complex DesK:DesR in Different Functional States Using Rational Mutagenesis and X-ray Crystallography.
Snapshots of the Signaling Complex DesK:DesR in Different Functional States Using Rational Mutagenesis and X-ray Crystallography.
Bio-protocol (2017-08-20)
Juan Andres Imelio, Nicole Larrieux, Ariel Edgardo Mechaly, Felipe Trajtenberg, Alejandro Buschiazzo
ABSTRACT
We have developed protocols to generate site-specific variants of the histidine-kinase DesK and its cognate response regulator DesR, conducive to trapping different signaling states of the proteins. Co-expression of both partners in E. coli, ensuring an excess of the regulator, was essential for soluble production of the DesK:DesR complexes and further purification. The 3D structures of the complex trapped in the phosphotransferase and in the phosphatase reaction steps, were solved by X-ray crystallography using molecular replacement. The solution was not trivial, and we found that in silico-generated models used as search probes, were instrumental to succeeding in placing a large portion of the complex in the asymmetric unit. Electron density maps were then clear enough to allow for manual model building attaining complete atomic models. These methods contribute to tackling a major challenge in the bacterial signaling field, namely obtaining stable kinase:regulator complexes, in distinct conformational states, amenable for high-resolution crystallographic studies.
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
β,γ-Methyleneadenosine 5′-triphosphate disodium salt, ≥95%, solid
Sigma-Aldrich
Yeast Extract, for use in microbial growth medium
Sigma-Aldrich
MES hydrate, ≥99.5% (titration)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Triton X-100, BioXtra
Sigma-Aldrich
Ampicillin sodium salt
Sigma-Aldrich
Lithium potassium acetyl phosphate, high-energy phosphate donor
Sigma-Aldrich
Adenosine 5′-triphosphate (ATP) disodium salt hydrate, vial of 30 mg
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, suitable for electrophoresis, ≥99.0%
Sigma-Aldrich
Lysozyme from chicken egg white, powder or granules, ≥39,000 units/mg protein
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%