Merck
  • A proopiomelanocortin-derived peptide sequence enhances plasma stability of peptide drugs.
A proopiomelanocortin-derived peptide sequence enhances plasma stability of peptide drugs.
FEBS letters (2020-06-09)
Karin Löw, Alexandre Roulin, Stefan Kunz
ABSTRACT
Bioactive peptide drugs hold promise for therapeutic application due to their high potency and selectivity but display short plasma half-life. Examination of selected naturally occurring peptide hormones derived from proteolytic cleavage of the proopiomelanocortin (POMC) precursor lead to the identification of significant plasma-stabilizing properties of a 12-amino acid serine-rich orphan sequence NSSSSGSSGAGQ in human γ3-melanocyte-stimulating hormone (MSH) that is homologous to previously discovered NSn GGH (n = 4-24) sequences in owls. Notably, transfer of this sequence to des-acetyl-α-MSH and the therapeutically relevant peptide hormones neurotensin and glucagon-like peptide-1 likewise enhance their plasma stability without affecting receptor signaling. The stabilizing effect of the sequence module is independent of plasma components, suggesting a direct effect in cis. This natural sequence module may provide a possible strategy to enhance plasma stability, complementing existing methods of chemical modification.
Sigma-Aldrich
Albumin from human serum, lyophilized powder, essentially protease free, ≥96% (agarose gel electrophoresis)