Merck
  • The nascent polypeptide in the 60S subunit determines the Rqc2-dependency of ribosomal quality control.
The nascent polypeptide in the 60S subunit determines the Rqc2-dependency of ribosomal quality control.
Nucleic acids research (2021-01-30)
Masato Mizuno, Shuhei Ebine, Okuto Shounai, Shizuka Nakajima, Shota Tomomatsu, Ken Ikeuchi, Yoshitaka Matsuo, Toshifumi Inada
ABSTRACT
Ribosome stalling at tandem CGA codons or poly(A) sequences activates quality controls for nascent polypeptides including ribosome-associated quality control (RQC) and no-go mRNA decay (NGD). In RQC pathway, Hel2-dependent uS10 ubiquitination and the RQC-trigger (RQT) complex are essential for subunit dissociation, and Ltn1-dependent ubiquitination of peptidyl-tRNA in the 60S subunit requires Rqc2. Here, we report that polytryptophan sequences induce Rqc2-independent RQC. More than 11 consecutive tryptophan residues induced RQC in a manner dependent on Hel2-mediated ribosome ubiquitination and the RQT complex. Polytryptophan sequence-mediated RQC was not coupled with CAT-tailing, and Rqc2 was not required for Ltn1-dependent degradation of the arrest products. Eight consecutive tryptophan residues located at the region proximal to the peptidyl transferase center in the ribosome tunnel inhibited CAT-tailing by tandem CGA codons. Polytryptophan sequences also induced Hel2-mediated canonical RQC-coupled NGD and RQC-uncoupled NGD outside the stalled ribosomes. We propose that poly-tryptophan sequences induce Rqc2-independent RQC, suggesting that CAT-tailing in the 60S subunit could be modulated by the polypeptide in the ribosome exit tunnel.
Product Description
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep
Roche
DIG Easy Hyb Granules, powder, suitable for hybridization
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Roche
Anti-HA-Peroxidase, High Affinity, from rat IgG1