- Preparation of yeast media.
Preparation of yeast media.
Current protocols in molecular biology (2008-02-12)
D A Treco, V Lundblad
PMID18265093
ABSTRACT
Preparation of sterile media of consistently high quality is essential for the genetic manipulation of yeast. Recipes for media needed in the protocols in this chapter are provided in this unit. Specific suppliers are recommended for specific ingredients.