Merck
  • Distinct pathways of homologous recombination controlled by the SWS1-SWSAP1-SPIDR complex.
Distinct pathways of homologous recombination controlled by the SWS1-SWSAP1-SPIDR complex.
Nature communications (2021-07-14)
Rohit Prakash, Thomas Sandoval, Florian Morati, Jennifer A Zagelbaum, Pei-Xin Lim, Travis White, Brett Taylor, Raymond Wang, Emilie C B Desclos, Meghan R Sullivan, Hayley L Rein, Kara A Bernstein, Przemek M Krawczyk, Jean Gautier, Mauro Modesti, Fabio Vanoli, Maria Jasin
ABSTRACT
Homology-directed repair (HDR), a critical DNA repair pathway in mammalian cells, is complex, leading to multiple outcomes with different impacts on genomic integrity. However, the factors that control these different outcomes are often not well understood. Here we show that SWS1-SWSAP1-SPIDR controls distinct types of HDR. Despite their requirement for stable assembly of RAD51 recombinase at DNA damage sites, these proteins are not essential for intra-chromosomal HDR, providing insight into why patients and mice with mutations are viable. However, SWS1-SWSAP1-SPIDR is critical for inter-homolog HDR, the first mitotic factor identified specifically for this function. Furthermore, SWS1-SWSAP1-SPIDR drives the high level of sister-chromatid exchange, promotes long-range loss of heterozygosity often involved with cancer initiation, and impels the poor growth of BLM helicase-deficient cells. The relevance of these genetic interactions is evident as SWSAP1 loss prolongs Blm-mutant embryo survival, suggesting a possible druggable target for the treatment of Bloom syndrome.
Sigma-Aldrich
Neocarzinostatin from Streptomyces carzinostaticus, ≥90% (SDS-PAGE), ~0.5 mg/mL
Sigma-Aldrich
Anti-Rad51 (Ab-1) Rabbit pAb, liquid, Calbiochem®
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Roche
Biotin-16-dUTP
Sigma-Aldrich
Doxycycline hyclate
Sigma-Aldrich
Mouse IgG−Agarose, (Suspension in 0.5 M NaCl containing preservative.)
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
ML216, ≥98% (HPLC)