Merck
  • Layer 4 Gates Plasticity in Visual Cortex Independent of a Canonical Microcircuit.
Layer 4 Gates Plasticity in Visual Cortex Independent of a Canonical Microcircuit.
Current biology : CB (2020-06-27)
Michael G Frantz, Emily C Crouse, Guela Sokhadze, Taruna Ikrar, Céleste-Élise Stephany, Collins Nguyen, Xiangmin Xu, Aaron W McGee
ABSTRACT
Disrupting binocular vision during a developmental critical period can yield enduring changes to ocular dominance (OD) in primary visual cortex (V1). Here we investigated how this experience-dependent plasticity is coordinated within the laminar circuitry of V1 by deleting separately in each cortical layer (L) a gene required to close the critical period, nogo-66 receptor (ngr1). Deleting ngr1 in excitatory neurons in L4, but not in L2/3, L5, or L6, prevented closure of the critical period, and adult mice remained sensitive to brief monocular deprivation. Intracortical disinhibition, but not thalamocortical disinhibition, accompanied this OD plasticity. Both juvenile wild-type mice and adult mice lacking ngr1 in L4 displayed OD plasticity that advanced more rapidly L4 than L2/3 or L5. Interestingly, blocking OD plasticity in L2/3 with the drug AM-251 did not impair OD plasticity in L5. We propose that L4 restricts disinhibition and gates OD plasticity independent of a canonical cortical microcircuit.
Sigma-Aldrich
QX-314, A quaternary lidocaine derivative, permanently charged, and lipophobic.
Pricing and availability is not currently available.
Sigma-Aldrich
Triton X-100, BioXtra
Pricing and availability is not currently available.
Sigma-Aldrich
REDExtract-N-Amp Tissue PCR Kit, sufficient for 10 reactions, sufficient for 100 reactions, sufficient for 1000 reactions, hotstart, dNTPs included
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium azide, BioXtra
Pricing and availability is not currently available.