Merck
FDA-approved drug labeling for the study of drug-induced liver injury.
Drug discovery today (2011-06-01)
Minjun Chen, Vikrant Vijay, Qiang Shi, Zhichao Liu, Hong Fang, Weida Tong
ABSTRACT
Drug-induced liver injury (DILI) is a leading cause of drugs failing during clinical trials and being withdrawn from the market. Comparative analysis of drugs based on their DILI potential is an effective approach to discover key DILI mechanisms and risk factors. However, assessing the DILI potential of a drug is a challenge with no existing consensus methods. We proposed a systematic classification scheme using FDA-approved drug labeling to assess the DILI potential of drugs, which yielded a benchmark dataset with 287 drugs representing a wide range of therapeutic categories and daily dosage amounts. The method is transparent and reproducible with a potential to serve as a common practice to study the DILI of marketed drugs for supporting drug discovery and biomarker development.
Product Description
Supelco
Cyclosporin A, VETRANAL®, analytical standard
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
Nicotinic acid, ≥99.5% (HPLC)
Supelco
Ibuprofen
Supelco
Nitrofurantoin, VETRANAL®, analytical standard
Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)
Supelco
Mebendazole, VETRANAL®, analytical standard
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Supelco
Prednisolone, VETRANAL®, analytical standard
Sigma-Aldrich
Ethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC/NT)
Sigma-Aldrich
2-Mercapto-1-methylimidazole, ≥99%
Sigma-Aldrich
Dianhydro-D-glucitol, 98%
Supelco
Ethanolamine, analytical standard
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Supelco
Warfarin, PESTANAL®, analytical standard
Supelco
Nicotinic acid, analytical standard
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Supelco
Mebendazole, analytical standard, ≥98% (HPLC)
Sigma-Aldrich
Cyclosporin A, BioReagent, from Tolypocladium inflatum, for molecular biology, ≥95%
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
Sigma-Aldrich
Mevinolin from Aspergillus sp., ≥98% (HPLC)
Sigma-Aldrich
Pyrazinecarboxamide
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Carbamazepine, powder
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Nicotinic acid, meets USP testing specifications
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Cytosine β-D-arabinofuranoside, crystalline, ≥90% (HPLC)
Sigma-Aldrich
Griseofulvin, from Penicillium griseofulvum, 97.0-102.0%