Merck
  • Design of benzimidazole- and benzoxazole-2-thione derivatives as inhibitors of bacterial hyaluronan lyase.
Design of benzimidazole- and benzoxazole-2-thione derivatives as inhibitors of bacterial hyaluronan lyase.
European journal of medicinal chemistry (2011-08-02)
Stephan Braun, Alexander Botzki, Sunnhild Salmen, Christian Textor, Günther Bernhardt, Stefan Dove, Armin Buschauer
ABSTRACT
Bacterial hyaluronan lyases (Hyal) degrade hyaluronan, an important component of the extracellular matrix, and are involved in microbial spread. Hyal inhibitors may serve as tools to study the role of the enzyme, its substrates and products in the course of bacterial infections. Moreover, such enzyme inhibitors are potential candidates for antibacterial combination therapy. Based on crystal structures of Streptococcus pneumoniae Hyal in complex with a hexasaccharide substrate and with different inhibitors, 1-acylated benzimidazole-2-thiones and benzoxazole-2-thiones were derived as new leads for the inhibition of Streptococcus agalactiae strain 4755 Hyal. Structure-based optimization led to N-(3-phenylpropionyl)benzoxazole-2-thione, one of the most potent compounds known to date (IC(50) values: 24 μM at pH 7.4, 15 μM at pH 5). Among the 27 new derivatives, other N-acylated benzimidazoles and benzoxazoles are just as active at pH 7.4, but not at pH 5. The results support a binding mode characterized by interactions with residues in the catalytic site and with a hydrophobic patch.
Sigma-Aldrich
2-Mercaptobenzoxazole, 95%
Sigma-Aldrich
18β-Glycyrrhetinic acid, 97%
Sigma-Aldrich
Ascorbic acid 6-palmitate, meets USP testing specifications
Sigma-Aldrich
Hyaluronidase from Streptomyces hyalurolyticus
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)