Merck
  • Cellular Trafficking of Glutathione Transferase M2-2 Between U373MG and SHSY-S7 Cells is Mediated by Exosomes.
Cellular Trafficking of Glutathione Transferase M2-2 Between U373MG and SHSY-S7 Cells is Mediated by Exosomes.
Neurotoxicity research (2021-02-09)
Raúl Valdes, Alicia Armijo, Patricia Muñoz, Kjell Hultenby, Andres Hagg, Jose Inzunza, Ivan Nalvarte, Mukesh Varshney, Bengt Mannervik, Juan Segura-Aguilar
ABSTRACT
The enzyme glutathione transferase M2-2, expressed in human astrocytes, increases its expression in the presence of aminochrome and catalyzes the conjugation of aminochrome, preventing its toxic effects. Secretion of the enzyme glutathione transferase M2-2 from U373MG cells, used as a cellular model for astrocytes, has been reported, and the enzyme is taken up by neuroblastoma SYSH-S7 cells and provide protection against aminochrome. The present study provides evidence that glutathione transferase M2-2 is released in exosomes from U373MG cells, thereby providing a means for intercellular transport of the enzyme. With particular relevance to Parkinson disease and other degenerative conditions, we propose a new mechanism by which astrocytes may protect dopaminergic neurons against the endogenous neurotoxin aminochrome.
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Amersham Hybond® LFP Western blotting membranes, PVDF, pore size 0.2 μm, roll W × L 254 mm × 4 m, pkg of 1 ea
Sigma-Aldrich
Dopamine hydrochloride