Merck
  • Deep eutectic solvent-based liquid-phase microextraction coupled with reversed-phase high-performance liquid chromatography for determination of α-, β-, γ-, and δ-tocopherol in edible oils.
Deep eutectic solvent-based liquid-phase microextraction coupled with reversed-phase high-performance liquid chromatography for determination of α-, β-, γ-, and δ-tocopherol in edible oils.
Analytical and bioanalytical chemistry (2020-11-19)
Qilong Xie, Ming Xia, Dekui Sun, Jiangping Cao, Yong Xiao, Mingui Lin, Bo Hou, Litao Jia, Debao Li
ABSTRACT
For simultaneous analysis of four fat-soluble tocopherols (α-, β-, γ-, and δ-) in edible oils, an efficient and green method using deep eutectic solvent-based liquid-phase microextraction (DES-LPME) coupled with reversed-phase high-performance liquid chromatography (RP-HPLC) was developed. The DESs formed by different quaternary ammonium salts and ethanol were used as the extractants. Tetrabutylammonium chloride (TBAC)-ethanol DES at a molar ratio of 1:2 achieved the best extraction efficiency. Under the optimized conditions, the detection limits were in the range of 2.1-3.0 ng mL-1. The intra-day and inter-day repeatability were in the ranges of 3.9-5.3% and 4.8-7.1%, respectively, and the recoveries for the real samples varied from 80.7% to 105.4%. The developed method was successfully employed for the determination of all four tocopherol homologues with an RP-HPLC system containing a COSMOSIL π-NAP column in five edible oils collected locally. Graphical abstract.
Sigma-Aldrich
Tetrapropylammonium chloride, 98%