Merck
  • miR‑335 promotes ferroptosis by targeting ferritin heavy chain 1 in in vivo and in vitro models of Parkinson's disease.
miR‑335 promotes ferroptosis by targeting ferritin heavy chain 1 in in vivo and in vitro models of Parkinson's disease.
International journal of molecular medicine (2021-03-03)
Xinrong Li, Wenwen Si, Zhan Li, Ye Tian, Xuelei Liu, Shanyu Ye, Zifeng Huang, Yichun Ji, Caiping Zhao, Xiaoqian Hao, Dongfeng Chen, Meiling Zhu
ABSTRACT
Parkinson's disease (PD) is a neurodegenerative disease characterized by the selective loss of dopaminergic neurons in the substantia nigra (SN). In a previous study, the authors demonstrated that ferritin heavy chain 1 (FTH1) inhibited ferroptosis in a model of 6‑hydroxydopamine (6‑OHDA)‑induced PD. However, whether and how microRNAs (miRNAs/miRs) modulate FTH1 in PD ferroptosis is not yet well understood. In the present study, in vivo and in vitro models of PD induced by 6‑OHDA were established. The results in vivo and in vitro revealed that the levels of the ferroptosis marker protein, glutathione peroxidase 4 (GPX4), and the PD marker protein, tyrosine hydroxylase (TH), were decreased in the model group, associated with a decreased FTH1 expression and the upregulation of miR‑335. In both the in vivo and in vitro models, miR‑335 mimic led to a lower FTH1 expression, exacerbated ferroptosis and an enhanced PD pathology. The luciferase 3'‑untranslated region reporter results identified FTH1 as the direct target of miR‑335. The silencing of FTH1 in 6‑OHDA‑stimulated cells enhanced the effects of miR‑335 on ferroptosis and promoted PD pathology. Mechanistically, miR‑335 enhanced ferroptosis through the degradation of FTH1 to increase iron release, lipid peroxidation and reactive oxygen species (ROS) accumulation, and to decrease mitochondrial membrane potential (MMP). On the whole, the findings of the present study reveal that miR‑335 promotes ferroptosis by targeting FTH1 in in vitro and in vivo models of PD, providing a potential therapeutic target for the treatment of PD.
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from rabbit
Sigma-Aldrich
6-Hydroxydopamine hydrochloride, ≥97% (titration), powder