Merck
  • HDAC inhibitors suppress the proliferation, migration and invasiveness of human head and neck squamous cell carcinoma cells via p63‑mediated tight junction molecules and p21‑mediated growth arrest.
HDAC inhibitors suppress the proliferation, migration and invasiveness of human head and neck squamous cell carcinoma cells via p63‑mediated tight junction molecules and p21‑mediated growth arrest.
Oncology reports (2021-03-03)
Akito Kakiuchi, Takuya Kakuki, Kizuku Ohwada, Makoto Kurose, Atsushi Kondoh, Kazufumi Obata, Kazuaki Nomura, Ryo Miyata, Yakuto Kaneko, Takumi Konno, Takayuki Kohno, Tetsuo Himi, Ken-Ichi Takano, Takashi Kojima
ABSTRACT
In human head and neck squamous cell carcinoma (HNSCC), the invasion and metastatic properties of cancer cells are promoted by junctional adhesion molecule‑A (JAM‑A) and claudin‑1; these are epithelial tight junction molecules regulated by histone deacetylases (HDACs) and transcription factor p63. HDAC expression is reportedly upregulated in HNSCC, and HDAC inhibitors suppress cancer cell proliferation by initiating proliferative arrest or apoptosis. However, little is known of the anti‑cancer mechanisms of HDAC inhibitors in HNSCC. Thus, in the present study, the HNSCC Detroit 562 cell line and primary cultured HNSCC cells were treated with HDAC inhibitors to investigate their effects in HNSCC. Higher expression of p63, HDAC1, JAM‑A and claudin‑1 was observed in HNSCC tissues compared with the adjacent dysplastic regions. In Detroit 562 cells, treatment with trichostatin A (TSA), an inhibitor of HDAC1 and 6, downregulated the expression of p63, JAM‑A and claudin‑1, and upregulated that of acetylated tubulin; conversely, p63 knockdown resulted in the downregulation of JAM‑A and claudin‑1. Collectively, inhibiting HDAC suppressed the migration and invasiveness of cancer cells. In addition, treatment with TSA suppressed cancer cell proliferation via G2/M arrest, as well as upregulating p21 and downregulating cyclin D1 expression. TSA also downregulated the expression of epidermal growth factor receptor (EGFR) and phospho‑ERK1/2. p63 knockdown and treatment with an EGFR inhibitor induced G1 arrest and downregulated EGFR and phospho‑ERK1/2 levels, respectively. HDAC inhibition also suppressed the migration and invasiveness of primary cultured HNSCC cells. Collectively, the results of the present study indicate that HDAC inhibitors suppress the proliferation, migration and invasiveness of HNSCC by downregulating the p63‑mediated tight junction molecules JAM‑A and claudin‑1, and inducing p63 or p21‑mediated growth arrest.
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
AG 1478, A cell-permeable, reversible, ATP-competitive, highly potent and selective inhibitor of epidermal growth factor receptor kinase versus HER2-neu and platelet-derived growth factor receptor kinase.
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.