Merck
  • Chromosomally encoded blaCMY-2 located on a novel SXT/R391-related integrating conjugative element in a Proteus mirabilis clinical isolate.
Chromosomally encoded blaCMY-2 located on a novel SXT/R391-related integrating conjugative element in a Proteus mirabilis clinical isolate.
Antimicrobial agents and chemotherapy (2010-06-23)
Sohei Harada, Yoshikazu Ishii, Tomoo Saga, Kazuhiro Tateda, Keizo Yamaguchi
ABSTRACT
Integrating conjugative elements (ICEs) are mobile genetic elements that can transfer from the chromosome of a host to the chromosome of a new host through the process of excision, conjugation, and integration. Although SXT/R391-related ICEs, originally demonstrated in Vibrio cholerae O139 isolates, have become prevalent among V. cholerae isolates in Asia, the prevalence of the ICEs among gram-negative bacteria other than Vibrio spp. remains unknown. In addition, SXT/R391-related ICEs carrying genes conferring resistance to extended-spectrum cephalosporins have never been described. Here we carried out a genetic analysis of a cefoxitin-resistant Proteus mirabilis clinical isolate, TUM4660, which revealed the presence of a novel SXT/R391-related ICE, ICEPmiJpn1. ICEPmiJpn1 had a core genetic structure showing high similarity to that of R391 and carried xis and int genes completely identical to those of R391, while an IS10-mediated composite transposon carrying bla(CMY-2) was integrated into the ICE. A nucleotide sequence identical to the 3' part of ISEcp1 was located upstream of the bla(CMY-2) gene, and other genes observed around bla(CMY-2) in earlier studies were also present. Furthermore, the nucleotide sequences of hot spot 2 and hot spot 4 in ICEPmiJpn1 showed high similarity to that of hot spot 2 in SXT(MO10) and with a part of the nucleotide sequence found in P. mirabilis ATCC 29906, respectively. ICEPmiJpn1 was successfully transferred to Escherichia coli, Klebsiella pneumoniae, Salmonella enterica serovar Typhimurium, and Citrobacter koseri in conjugation experiments. These observations suggest that ICEs may contribute to the dissemination of antimicrobial resistance genes among clinically relevant Enterobacteriaceae, which warrants careful observation of the prevalence of ICEs, including SXT/R391-related ICEs.
Sigma-Aldrich
Trimethoprim, ≥98.5%
Supelco
Trimethoprim, VETRANAL®, analytical standard
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Amoxicillin, 95.0-102.0% anhydrous basis