Merck
  • The Oncogenic PRL Protein Causes Acid Addiction of Cells by Stimulating Lysosomal Exocytosis.
The Oncogenic PRL Protein Causes Acid Addiction of Cells by Stimulating Lysosomal Exocytosis.
Developmental cell (2020-09-13)
Yosuke Funato, Atsushi Yoshida, Yusuke Hirata, Osamu Hashizume, Daisuke Yamazaki, Hiroaki Miki
ABSTRACT
Extracellular pH is usually maintained around 7.4 in multicellular organisms, and cells are optimized to proliferate under this condition. Here, we find cells can adapt to a more acidic pH of 6.5 and become addicted to this acidic microenvironment by expressing phosphatase of regenerating liver (PRL), a driver of cancer malignancy. Genome-scale CRISPR-Cas9 knockout screening and subsequent analyses revealed that PRL promotes H+ extrusion and acid addiction by stimulating lysosomal exocytosis. Further experiments using cultured cells and Caenorhabditis elegans clarified the molecular link between PRL and lysosomal exocytosis across species, involving activation of lysosomal Ca2+ channel TRPML by ROS. Indeed, disruption of TRPML in cancer cells abolished PRL-stimulated lysosomal exocytosis, acid addiction, and metastasis. Thus, PRL is the molecular switch turning cells addicted to an acidic condition, which should benefit cancer cells to thrive in an acidic tumor microenvironment.
Sigma-Aldrich
5-(N-Ethyl-N-isopropyl)amiloride
Sigma-Aldrich
Anti-Myosin IIA, non muscle antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
4-Nitrophenyl N-acetyl-β-D-glucosaminide, ≥99% (TLC)
Sigma-Aldrich
Calcium propionate, ≥97.0% (dry substance, NT)
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Dimethyl pimelimidate dihydrochloride, powder
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Supelco
2,3-Butanedione, analytical standard
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Porapak Porous Polymer Adsorbent, matrix Porapak R, 50-80 mesh, bottle of 75 cc