Merck
  • Evaluation of anti-pigmentary effect of synthetic sulfonylamino chalcone.
Evaluation of anti-pigmentary effect of synthetic sulfonylamino chalcone.
European journal of medicinal chemistry (2010-02-13)
Woo Duck Seo, Young Bae Ryu, Marcus J Curtis-Long, Chan Woo Lee, Hyung Won Ryu, Ki Chang Jang, Ki Hun Park
ABSTRACT
The 4'-(p-toluenesulfonylamino)-4-hydroxychalcone (TSAHC), which bears inhibitory chemotypes for both alpha-glucosidase and tyrosinase, was evaluated for tyrosinase activity and depigmenting ability relative to compounds designed to only target tyrosianse activity. TSAHC emerged to be a competitive reversible inhibitor of mushroom tyrosinase. More importantly, it was also able to return the melanin content of alpha-melanocyte stimulated by alpha-MSH to base levels unlike other inhibitors that only targeted tyrosinase. The Western blot for expression levels of proteins involved in melanogenesis showed that TSAHC significantly decreased three main tyrosinase related protein in melanin biosynthesis, tyrosinase, TRP-1 and TRP-2.
Sigma-Aldrich
Hydroquinone, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydroquinone, meets USP testing specifications
Sigma-Aldrich
Kojic acid
Sigma-Aldrich
Hydroquinone, ReagentPlus®, 99%