Merck
  • Targeted epigenetic modulation using a DNA-based histone deacetylase inhibitor enhances cardiomyogenesis in mouse embryonic stem cells.
Targeted epigenetic modulation using a DNA-based histone deacetylase inhibitor enhances cardiomyogenesis in mouse embryonic stem cells.
Journal of cellular physiology (2020-11-10)
Jin-A Lee, Jieun An, Junichi Taniguchi, Gengo Kashiwazaki, Ganesh N Pandian, Nazia Parveen, Tong Mook Kang, Hiroshi Sugiyama, Debojyoti De, Kyeong Kyu Kim
ABSTRACT
The epigenome has an essential role in orchestrating transcriptional activation and modulating key developmental processes. Previously, we developed a library of pyrrole-imidazole polyamides (PIPs) conjugated with suberoylanilide hydroxamic acid (SAHA), a histone deacetylase (HDAC) inhibitor, for the purpose of sequence-specific modification of epigenetics. Based on the gene expression profile of SAHA-PIPs and screening studies using the α-myosin heavy chain promoter-driven reporter and SAHA-PIP library, we identified that SAHA-PIP G activates cardiac-related genes. Studies in mouse ES cells showed that SAHA-PIP G could enhance the generation of spontaneous beating cells, which is consistent with upregulation of several cardiac-related genes. Moreover, ChIP-seq results confirmed that the upregulation of cardiac-related genes is highly correlated with epigenetic activation, relevant to the sequence-specific binding of SAHA-PIP G. This proof-of-concept study demonstrating the applicability of SAHA-PIP not only improves our understanding of epigenetic alterations involved in cardiomyogenesis but also provides a novel chemical-based strategy for stem cell differentiation.
Sigma-Aldrich
Anti-α-Actinin (Sarcomeric) antibody, Mouse monoclonal, clone EA-53, purified from hybridoma cell culture
Corning® Costar® Ultra-Low Attachment Multiple Well Plate, size 24 well, flat bottom clear, pkg of (individually wrapped), pkg of (24/case), sterile, lid
Sigma-Aldrich
DAPI, for nucleic acid staining
Swagelok® Union Cross, Swagelok® 400-4, brass, 1/4 in. Swagelok
Sigma-Aldrich
Blasticidine S hydrochloride
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%