Merck
  • The Orphan Nuclear Receptor TLX Is a Receptor for Synthetic and Natural Retinoids.
The Orphan Nuclear Receptor TLX Is a Receptor for Synthetic and Natural Retinoids.
Cell chemical biology (2020-08-09)
Kristine Griffett, Gonzalo Bedia-Diaz, Lamees Hegazy, Ian Mitchelle S de Vera, Udayanga S Wanninayake, Cyrielle Billon, Thomas Koelblen, McKenna L Wilhelm, Thomas P Burris
ABSTRACT
TLX is an orphan nuclear receptor that plays a critical role in both embryonic and adult neurogenesis, as well in the pathogenesis of glioblastomas. TLX functions predominately as a transcriptional repressor, but no natural ligands or high-affinity synthetic ligands have been identified. Here, we describe the identification of natural and synthetic retinoids as functional ligands for TLX. We identified potent synthetic retinoids that directly bind to TLX and either activate or inhibit its transcriptional repressor activity. Furthermore, we identified all-trans and 11-cis retinaldehyde (retinal), retinoids that play an essential role in the visual cycle, as the preferential natural retinoids that bind to and modulate the function of TLX. Molecular dynamics simulations followed by mutational analysis provided insight into the molecular basis of retinoid binding to TLX. Our data support the validity of TLX as a target for development of therapeutics to treat cognitive disorders and/or glioblastomas.
Sigma-Aldrich
Retinol, synthetic, ≥95% (HPLC), (Powder or Powder with Lumps)
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting human NR2E1
Sigma-Aldrich
all trans-Retinal, powder, ≥98%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Nitrilotriacetic acid disodium salt, Sigma Grade, ≥99%