Merck
  • Involvement of PP2A methylation in the adipogenic differentiation of bone marrow-derived mesenchymal stem cell.
Involvement of PP2A methylation in the adipogenic differentiation of bone marrow-derived mesenchymal stem cell.
Journal of biochemistry (2020-07-15)
Shunta Ikeda, Shunya Tsuji, Takashi Ohama, Koichi Sato
ABSTRACT
Bone marrow-derived mesenchymal stem cells (BM-MSCs) are multipotent stem cells with ability to self-replicate and differentiate into mesodermal derivatives, such as adipocytes and osteoblasts. BM-MSCs are a critical component of the tumour microenvironment. They support tumour progression by recruiting additional BM-MSCs and by differentiating into myofibroblasts (also called cancer-associated fibroblasts). Protein phosphatase 2A (PP2A) is an essential serine/threonine protein phosphatase that regulates a broad range of cellular signalling. PP2A forms a heterotrimer to dephosphorylate specific substrates. The reversible methylesterification (methylation) of Leu309 in the catalytic subunit of PP2A (PP2Ac) regulates biogenesis of the PP2A holoenzyme. It is unknown whether the methylation of PP2Ac plays a role in BM-MSC differentiation. Our experiments determined that protein levels of PP2A subunits and PP2A methyltransferase (LCMT-1) are significantly altered during differentiation. PP2Ac methylation levels in BM-MSCs decrease over time in response to an adipogenic differentiation stimulus. However, blockage of PP2A demethylation using the PP2A dimethyl-esterase inhibitors enhanced adipocyte differentiation. This suggests that PP2Ac demethylation is involved in adipocyte differentiation resistance. The results of our study provide a greater understanding of the regulation of BM-MSCs differentiation by PP2A holoenzyme.
Sigma-Aldrich
ABL127, ≥98% (HPLC)