Merck
  • Simultaneous cortisol/insulin microchip detection using dual enzyme tagging.
Simultaneous cortisol/insulin microchip detection using dual enzyme tagging.
Biosensors & bioelectronics (2020-09-03)
Eva Vargas, Eloy Povedano, Sadagopan Krishnan, Hazhir Teymourian, Farshad Tehrani, Susana Campuzano, Eyal Dassau, Joseph Wang
ABSTRACT
Here we describe the development of a dual electrochemical immunosensor microchip for simultaneous detection of insulin (I) and cortisol (C) biomarkers that can enhance the ability to improve glucose regulation using automated insulin delivery. The successful realization of the simultaneous I and C measurements has been realized by integrating different enzymatically-tagged competitive and sandwich immunoassay formats on a single chip platform. The insulin detection is based on a peroxidase (HRP)-labeled sandwich assay whereas the cortisol detection relies on an alkaline phosphatase (ALP)-labeled competitive immunoassay. The attractive analytical performance of the dual marker immunosensor, with no apparent cross-talk, was achieved through systematic optimization of the incubation and amperometric detection of the different captured enzyme tags. Evaluation of dual biosensor chip in untreated serum samples indicated favorable simultaneous detection of picomolar (pM) insulin and nanomolar (nM) cortisol concentrations in a single microliter sample droplet within less than 25min. The new dual immunosensor chip offers considerable promise for frequent decentralized testing of I and C towards a tighter glycemic control and improved management of diabetes.
Sigma-Aldrich
1-Naphthyl phosphate, 99%