Merck
  • Activity of the SNARE Protein SNAP29 at the Endoplasmic Reticulum and Golgi Apparatus.
Activity of the SNARE Protein SNAP29 at the Endoplasmic Reticulum and Golgi Apparatus.
Frontiers in cell and developmental biology (2021-03-16)
Elena Morelli, Elisa A Speranza, Enrica Pellegrino, Galina V Beznoussenko, Francesca Carminati, Massimiliano Garré, Alexander A Mironov, Marco Onorati, Thomas Vaccari
ABSTRACT
Snap29 is a conserved regulator of membrane fusion essential to complete autophagy and to support other cellular processes, including cell division. In humans, inactivating SNAP29 mutations causes CEDNIK syndrome, a rare multi-systemic disorder characterized by congenital neuro-cutaneous alterations. The fibroblasts of CEDNIK patients show alterations of the Golgi apparatus (GA). However, whether and how Snap29 acts at the GA is unclear. Here we investigate SNAP29 function at the GA and endoplasmic reticulum (ER). As part of the elongated structures in proximity to these membrane compartments, a pool of SNAP29 forms a complex with Syntaxin18, or with Syntaxin5, which we find is required to engage SEC22B-loaded vesicles. Consistent with this, in HeLa cells, in neuroepithelial stem cells, and in vivo, decreased SNAP29 activity alters GA architecture and reduces ER to GA trafficking. Our data reveal a new regulatory function of Snap29 in promoting secretory trafficking.
Sigma-Aldrich
MISSION® esiRNA, targeting human SNAP29
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Insulin solution human, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human STX18
Sigma-Aldrich
Poly-L-ornithine solution, mol wt 30,000-70,000, 0.01%, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human STX5