Merck
  • Role of the Mycobacterium tuberculosis P55 efflux pump in intrinsic drug resistance, oxidative stress responses, and growth.
Role of the Mycobacterium tuberculosis P55 efflux pump in intrinsic drug resistance, oxidative stress responses, and growth.
Antimicrobial agents and chemotherapy (2009-07-01)
Santiago Ramón-García, Carlos Martín, Charles J Thompson, José A Aínsa
ABSTRACT
Bacterial efflux pumps have traditionally been studied as low-level drug resistance determinants. Recent insights have suggested that efflux systems are often involved with fundamental cellular physiological processes, suggesting that drug extrusion may be a secondary function. In Mycobacterium tuberculosis, little is known about the physiological or drug resistance roles of efflux pumps. Using Mycobacterium bovis BCG as a model system, we showed that deletion of the Rv1410c gene encoding the P55 efflux pump made the strain more susceptible to a range of toxic compounds, including rifampin (rifampicin) and clofazimine, which are first- and second-line antituberculosis drugs. The efflux pump inhibitors carbonyl cyanide m-chlorophenylhydrazone (CCCP) and valinomycin inhibited the P55-determined drug resistance, suggesting the active export of the compounds by use of the transmembrane proton and electrochemical gradients as sources of energy. In addition, the P55 efflux pump mutant was more susceptible to redox compounds and displayed increased intracellular redox potential, suggesting an essential role of the efflux pump in detoxification processes coupled to oxidative balance within the cell. Finally, cells that lacked the p55 gene displayed smaller colony sizes and had a growth defect in liquid culture. This, together with an increased susceptibility to the cell wall-targeting compounds bacitracin and vancomycin, suggested that P55 is needed for proper cell wall assembly and normal growth in vitro. Thus, P55 plays a fundamental role in oxidative stress responses and in vitro cell growth, in addition to contributing to intrinsic antibiotic resistance. Inhibitors of the P55 efflux pump could help to improve current treatments for tuberculosis.
Product Description
Supelco
Menadione (K3), analytical standard
Sigma-Aldrich
Menadione, crystalline
Sigma-Aldrich
Menadione, meets USP testing specifications
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Sigma-Aldrich
Reserpine
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Supelco
Reserpine Standard for LC-MS, analytical standard, for LC-MS
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Bromobimane, ≥97% (HPLC)