Merck
  • Single-Molecule 3D Orientation Imaging Reveals Nanoscale Compositional Heterogeneity in Lipid Membranes.
Single-Molecule 3D Orientation Imaging Reveals Nanoscale Compositional Heterogeneity in Lipid Membranes.
Angewandte Chemie (International ed. in English) (2020-07-11)
Jin Lu, Hesam Mazidi, Tianben Ding, Oumeng Zhang, Matthew D Lew
ABSTRACT
In soft matter, thermal energy causes molecules to continuously translate and rotate, even in crowded environments, thereby impacting the spatial organization and function of most molecular assemblies, such as lipid membranes. Directly measuring the orientation and spatial organization of large collections (>3000 molecules μm-2 ) of single molecules with nanoscale resolution remains elusive. In this paper, we utilize SMOLM, single-molecule orientation localization microscopy, to directly measure the orientation spectra (3D orientation plus "wobble") of lipophilic probes transiently bound to lipid membranes, revealing that Nile red's (NR) orientation spectra are extremely sensitive to membrane chemical composition. SMOLM images resolve nanodomains and enzyme-induced compositional heterogeneity within membranes, where NR within liquid-ordered vs. liquid-disordered domains shows a ≈4° difference in polar angle and a ≈0.3π sr difference in wobble angle. As a new type of imaging spectroscopy, SMOLM exposes the organizational and functional dynamics of lipid-lipid, lipid-protein, and lipid-dye interactions with single-molecule, nanoscale resolution.
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Glucose Oxidase from Aspergillus niger, Type VII, lyophilized powder, ≥100,000 units/g solid (without added oxygen)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
N-Palmitoyl-D-sphingomyelin, ≥96.0% (TLC)
Sigma-Aldrich
Melatonin, powder, ≥98% (TLC)
Sigma-Aldrich
Sphingomyelinase from Bacillus cereus, lyophilized powder, ≥100 units/mg protein
Sigma-Aldrich
Cholesterol-Water Soluble, powder, BioReagent, suitable for cell culture