Merck
  • Ciliopathy protein HYLS1 coordinates the biogenesis and signaling of primary cilia by activating the ciliary lipid kinase PIPKIγ.
Ciliopathy protein HYLS1 coordinates the biogenesis and signaling of primary cilia by activating the ciliary lipid kinase PIPKIγ.
Science advances (2021-06-25)
Chuan Chen, Qingwen Xu, Yuxia Zhang, Brian A Davies, Yan Huang, David J Katzmann, Peter C Harris, Jinghua Hu, Kun Ling
ABSTRACT
Mutation of ciliopathy protein HYLS1 causes the perinatal lethal hydrolethalus syndrome (HLS), yet the underlying molecular etiology and pathogenesis remain elusive. Here, we reveal unexpected mechanistic insights into the role of mammalian HYLS1 in regulating primary cilia. HYLS1 is recruited to the ciliary base via a direct interaction with the type Iγ phosphatidylinositol 4-phosphate [PI(4)P] 5-kinase (PIPKIγ). HYLS1 activates PIPKIγ by interrupting the autoinhibitory dimerization of PIPKIγ, which thereby expedites depletion of centrosomal PI(4)P to allow axoneme nucleation. HYLS1 deficiency interrupts the assembly of ciliary NPHP module and agonist-induced ciliary exit of β-arrestin, which, in turn, disturbs the removal of ciliary Gpr161 and activation of hedgehog (Hh) signaling. Consistent with this model of pathogenesis, the HLS mutant HYLS1D211G supports ciliogenesis but not activation of Hh signaling. These results implicate mammalian HYLS1 as a multitasking protein that facilitates ciliogenesis and ciliary signaling by coordinating with the ciliary lipid kinase PIPKIγ.
Sigma-Aldrich
Anti-TTBK2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Smoothened Agonist, SAG, A cell-permeable Smoothened Agonist, SAG, CAS 364590-63-6, modulates the coupling of Smo with its downstream effector by interacting with the Smo heptahelical domain (KD = 59 nM).
Sigma-Aldrich
Anti-HA antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture
Sigma-Aldrich
Anti-CEP83 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid