Merck
  • Protocol for Single-Nucleus Transcriptomics of Diploid and Tetraploid Cardiomyocytes in Murine Hearts.
Protocol for Single-Nucleus Transcriptomics of Diploid and Tetraploid Cardiomyocytes in Murine Hearts.
STAR protocols (2020-10-29)
Miao Cui, Eric N Olson
ABSTRACT
Murine cardiomyocytes undergo proliferation, multinucleation, and polyploidization during the first 3 weeks of postnatal life, resulting in a mixture of diploid and tetraploid cardiomyocytes in the heart. Understanding the molecular differences between diploid and tetraploid cardiomyocytes from these processes has been limited due to lack of unique markers and their heterogenous origins. Here, we apply single-nucleus RNA-sequencing to fluorescence-activated cell sorting-selected diploid and tetraploid cardiomyocytes to characterize their heterogeneity and molecular distinctions. For complete details on the use and execution of this protocol, please refer to Cui et al. (2020).
Sigma-Aldrich
Anti-PCM1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
Sigma-Aldrich
Spermidine trihydrochloride, ≥98% (TLC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)