Merck
Proteomic Analysis of Unbounded Cellular Compartments: Synaptic Clefts.
Cell (2016-08-28)
Ken H Loh, Philipp S Stawski, Austin S Draycott, Namrata D Udeshi, Emily K Lehrman, Daniel K Wilton, Tanya Svinkina, Thomas J Deerinck, Mark H Ellisman, Beth Stevens, Steven A Carr, Alice Y Ting
ABSTRACT
Cellular compartments that cannot be biochemically isolated are challenging to characterize. Here we demonstrate the proteomic characterization of the synaptic clefts that exist at both excitatory and inhibitory synapses. Normal brain function relies on the careful balance of these opposing neural connections, and understanding how this balance is achieved relies on knowledge of their protein compositions. Using a spatially restricted enzymatic tagging strategy, we mapped the proteomes of two of the most common excitatory and inhibitory synaptic clefts in living neurons. These proteomes reveal dozens of synaptic candidates and assign numerous known synaptic proteins to a specific cleft type. The molecular differentiation of each cleft allowed us to identify Mdga2 as a potential specificity factor influencing Neuroligin-2's recruitment of presynaptic neurotransmitters at inhibitory synapses.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
iTRAQ® Reagents Multiplex Kit
Sigma-Aldrich
Monoclonal Anti-Synaptophysin antibody produced in mouse, clone SVP-38, ascites fluid
Sigma-Aldrich
Biotin-XX-tyramide, ≥95% (HPLC)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use in purification of Histidine-tagged proteins, DMSO solution