Merck
  • ERK phosphorylates chromosomal axis component HORMA domain protein HTP-1 to regulate oocyte numbers.
ERK phosphorylates chromosomal axis component HORMA domain protein HTP-1 to regulate oocyte numbers.
Science advances (2020-11-01)
Debabrata Das, Shin-Yu Chen, Swathi Arur
ABSTRACT
Oocyte numbers, a critical determinant of female reproductive fitness, are highly regulated, yet the mechanisms underlying this regulation remain largely undefined. In the Caenorhabditis elegans gonad, RAS/extracellular signal-regulated kinase (ERK) signaling regulates oocyte numbers; mechanisms are unknown. We show that the RAS/ERK pathway phosphorylates meiotic chromosome axis protein HTP-1 at serine-325 to control chromosome dynamics and regulate oocyte number. Phosphorylated HTP-1(S325) accumulates in vivo in an ERK-dependent manner in early-mid pachytene stage germ cells and is necessary for synaptonemal complex extension and/or maintenance. Lack of HTP-1 phosphorylation leads to asynapsis and persistence of meiotic double-strand breaks, causing delayed meiotic progression and reduced oocyte number. In contrast, early onset of ERK activation causes precocious meiotic progression, resulting in increased oocyte number, which is reversed by removal of HTP-1 phosphorylation. The RAS/ERK/HTP-1 signaling cascade thus functions to monitor formation and maintenance of synapsis for timely resolution of double-strand breaks, oocyte production, and reproductive fitness.
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, clone 3H10, clone 3H10, Upstate®, from mouse
Sigma-Aldrich
Monoclonal Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2) antibody produced in mouse, clone MAPK-YT, ascites fluid
Sigma-Aldrich
Monoclonal Anti-polyHistidine antibody produced in mouse, clone HIS-1, ascites fluid
Sigma-Aldrich
BL21(DE3)pLysS Competent Cells - Novagen, BL21 host strain that expresses T7 RNA polymerase and also encode T7 lysozyme that suppresses basal expression of toxic target proteins prior to induction.