Merck
  • Constitutive aryl hydrocarbon receptor signaling constrains type I interferon-mediated antiviral innate defense.
Constitutive aryl hydrocarbon receptor signaling constrains type I interferon-mediated antiviral innate defense.
Nature immunology (2016-04-19)
Taisho Yamada, Hiromasa Horimoto, Takeshi Kameyama, Sumio Hayakawa, Hiroaki Yamato, Masayoshi Dazai, Ayato Takada, Hiroshi Kida, Debbie Bott, Angela C Zhou, David Hutin, Tania H Watts, Masahiro Asaka, Jason Matthews, Akinori Takaoka
ABSTRACT
Aryl hydrocarbon receptor (AHR) is a ligand-activated transcription factor that mediates the toxic activity of many environmental xenobiotics. However, its role in innate immune responses during viral infection is not fully understood. Here we demonstrate that constitutive AHR signaling negatively regulates the type I interferon (IFN-I) response during infection with various types of virus. Virus-induced IFN-β production was enhanced in AHR-deficient cells and mice and resulted in restricted viral replication. We found that AHR upregulates expression of the ADP-ribosylase TIPARP, which in turn causes downregulation of the IFN-I response. Mechanistically, TIPARP interacted with the kinase TBK1 and suppressed its activity by ADP-ribosylation. Thus, this study reveals the physiological importance of endogenous activation of AHR signaling in shaping the IFN-I-mediated innate response and, further, suggests that the AHR-TIPARP axis is a potential therapeutic target for enhancing antiviral responses.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-TIPARP antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid