Merck
  • A peptide tag-specific nanobody enables high-quality labeling for dSTORM imaging.
A peptide tag-specific nanobody enables high-quality labeling for dSTORM imaging.
Nature communications (2018-03-04)
David Virant, Bjoern Traenkle, Julia Maier, Philipp D Kaiser, Mona Bodenhöfer, Christian Schmees, Ilijana Vojnovic, Borbála Pisak-Lukáts, Ulrike Endesfelder, Ulrich Rothbauer
ABSTRACT
Dense fluorophore labeling without compromising the biological target is crucial for genuine super-resolution microscopy. Here we introduce a broadly applicable labeling strategy for fixed and living cells utilizing a short peptide tag-specific nanobody (BC2-tag/bivBC2-Nb). BC2-tagging of ectopically introduced or endogenous proteins does not interfere with the examined structures and bivBC2-Nb staining results in a close-grained fluorophore labeling with minimal linkage errors. This allowed us to perform high-quality dSTORM imaging of various targets in mammalian and yeast cells. We expect that this versatile strategy will render many more demanding cellular targets amenable to dSTORM imaging.
Sigma-Aldrich
Methyl viologen dichloride hydrate, 98%
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture