Merck
  • Polyethylene glycol improves current methods for circulating extracellular vesicle-derived DNA isolation.
Polyethylene glycol improves current methods for circulating extracellular vesicle-derived DNA isolation.
Journal of translational medicine (2019-03-16)
N García-Romero, R Madurga, G Rackov, I Palacín-Aliana, R Núñez-Torres, A Asensi-Puig, J Carrión-Navarro, S Esteban-Rubio, H Peinado, A González-Neira, V González-Rumayor, C Belda-Iniesta, A Ayuso-Sacido
ABSTRACT
Extracellular vesicles (EVs) are small membrane-bound vesicles which play an important role in cell-to-cell communication. Their molecular cargo analysis is presented as a new source for biomarker detection, and it might provide an alternative to traditional solid biopsies. However, the most effective approach for EV isolation is not yet well established. Here, we study the efficiency of the most common EV isolation methods-ultracentrifugation, Polyethlyene glycol and two commercial kits, Exoquick® and PureExo®. We isolated circulating EVs from the bloodstream of healthy donors, characterized the size and yield of EVs and analyzed their protein profiles and concentration. Moreover, we have used for the first time Digital-PCR to identify and detect specific gDNA sequences, which has several implications for diagnostic and monitoring many types of diseases. Our findings present Polyethylene glycol precipitation as the most feasible and less cost-consuming EV isolation technique.
Sigma-Aldrich
Proteinase K from Tritirachium album, buffered aqueous glycerol solution, for molecular biology, ≥800 units/mL
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Transcreener® AMP2/GMP2 FP Assay