Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Supelco
Cycloheximide, PESTANAL®, analytical standard
Supelco
Trimethoprim, VETRANAL®, analytical standard
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Sigma-Aldrich
Acridine Orange solution, 2% in H2O
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Gentian Violet, meets USP testing specifications
Sigma-Aldrich
Trimethoprim, ≥98.5%
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Supelco
Sulfathiazole, VETRANAL®, analytical standard
Sigma-Aldrich
Gentian violet, for microscopy (Bact., Hist.)
Sigma-Aldrich
Crystal Violet, S. No.: 785
Sigma-Aldrich
Crystal Violet, certified by the Biological Stain Commission
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Sigma-Aldrich
Fusaric acid, from Gibberella fujikuroi
Sigma-Aldrich
Nalidixic acid, ≥98%
Sigma-Aldrich
Crystal Violet Solution
Sigma-Aldrich
Crystal Violet Solution
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Ethidium bromide, BioReagent, for molecular biology, powder
Sigma-Aldrich
L-(−)-Dithiothreitol, ≥95% (titration)
Supelco
Crystal Violet, VETRANAL®, analytical standard
Sigma-Aldrich
Crystal Violet, for microscopy (Bact., Bot., Hist., Vit.), indicator (pH 0.1-2.0)