Merck
ISEcp1-mediated transposition of qnrB-like gene in Escherichia coli.
Antimicrobial agents and chemotherapy (2008-06-04)
Vincent Cattoir, Patrice Nordmann, Jesus Silva-Sanchez, Paula Espinal, Laurent Poirel
ABSTRACT
A novel QnrB-like plasmid-mediated resistance determinant, QnrB19, was identified from an Escherichia coli clinical isolate from Colombia. Its gene was associated with an ISEcp1-like insertion element that did not act as a promoter for its expression. Using an in vitro model of transposition, we showed that the ISEcp1-like element was able to mobilize the qnrB19 gene.
Product Description
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Supelco
Enrofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Enrofloxacin, ≥99.0%
Sigma-Aldrich
Sparfloxacin, ≥98.0% (HPLC)
Supelco
Norfloxacin, analytical standard, ≥98% (TLC)
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Sigma-Aldrich
Nalidixic acid, ≥98%
Supelco
Norfloxacin, VETRANAL®, analytical standard
Supelco
Ofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)