Merck
  • Biophysical stimulation induces demyelination via an integrin-dependent mechanism.
Biophysical stimulation induces demyelination via an integrin-dependent mechanism.
Annals of neurology (2012-07-26)
Michael Y Lin, Laura S Frieboes, Maryam Forootan, Winnie A Palispis, Tahseen Mozaffar, Matiar Jafari, Oswald Steward, Christine M Gall, Ranjan Gupta
ABSTRACT
Chronic nerve compression (CNC) injuries occur when peripheral nerves are subjected to sustained mechanical forces, with increasing evidence implicating Schwann cells as key mediators. Integrins, a family of transmembrane adhesion molecules that are capable of intracellular signaling, have been implicated in a variety of biological processes such as myelination and nerve regeneration. In this study, we seek to define the physical stimuli mediating demyelination and to determine whether integrin plays a role in the demyelinating response. We used a previously described in vitro model of CNC injury where myelinating neuron-Schwann cell cocultures were subjected to independent manipulations of hydrostatic pressure, hypoxia, and glucose deprivation in a custom bioreactor. We assessed whether demyelination increased in response to applied manipulation and determined whether integrin-associated signaling cascades are upregulated. Biophysical stimulation of neural tissue induced demyelination and Schwann cell proliferation without neuronal or glial cytotoxicity or apoptosis. Although glucose deprivation and hypoxia independently had minor effects on myelin stability, together they potentiated the demyelinating effects of hydrostatic compression, and in combination, significantly destabilized myelin. Biophysical stimuli transiently increased phosphorylation of the integrin-associated tyrosine kinase Src within Schwann cells. Silencing this integrin signaling cascade blocked Src activation and prevented pressure-induced demyelination. Colocalization analysis indicated that Src is localized within Schwann cells. These results indicate that myelin is sensitive to CNC injury and support the novel concept that myelinating cocultures respond directly to mechanical loading via activating an integrin signaling cascade.
Sigma-Aldrich
Donkey Anti-Mouse IgG Antibody, HRP conjugate, Species Adsorbed, Chemicon®, from donkey
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mg
1 Estimated to ship on 09 March 2025
CHF 34.30
1 g
1 Estimated to ship on 09 March 2025
CHF 137.00
100 g
1 Estimated to ship on 09 March 2025
CHF 1,230.00
500 g
1 Estimated to ship on 09 March 2025
CHF 2,140.00
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, Peroxidase Conjugated, 1 mg/mL (after reconstitution), Chemicon®
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mg
1 Estimated to ship on 09 March 2025
CHF 34.30
1 g
1 Estimated to ship on 09 March 2025
CHF 137.00
100 g
1 Estimated to ship on 09 March 2025
CHF 1,230.00
500 g
1 Estimated to ship on 09 March 2025
CHF 2,140.00
Sigma-Aldrich
Anti-Integrin β4 Antibody, clone ASC-3, azide free, clone ASC-3, Chemicon®, from mouse
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mg
1 Estimated to ship on 09 March 2025
CHF 34.30
1 g
1 Estimated to ship on 09 March 2025
CHF 137.00
100 g
1 Estimated to ship on 09 March 2025
CHF 1,230.00
500 g
1 Estimated to ship on 09 March 2025
CHF 2,140.00
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mg
1 Estimated to ship on 09 March 2025
CHF 34.30
1 g
1 Estimated to ship on 09 March 2025
CHF 137.00
100 g
1 Estimated to ship on 09 March 2025
CHF 1,230.00
500 g
1 Estimated to ship on 09 March 2025
CHF 2,140.00
Sigma-Aldrich
Anti-Actin (20-33) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
250 mg
1 Estimated to ship on 09 March 2025
CHF 34.30
1 g
1 Estimated to ship on 09 March 2025
CHF 137.00
100 g
1 Estimated to ship on 09 March 2025
CHF 1,230.00
500 g
1 Estimated to ship on 09 March 2025
CHF 2,140.00