Merck
  • Cytocompatible Anti-microbial Dressings of Syzygium cumini Cellulose Nanocrystals Decorated with Silver Nanoparticles Accelerate Acute and Diabetic Wound Healing.
Cytocompatible Anti-microbial Dressings of Syzygium cumini Cellulose Nanocrystals Decorated with Silver Nanoparticles Accelerate Acute and Diabetic Wound Healing.
Scientific reports (2017-09-07)
Rubbel Singla, Sourabh Soni, Vikram Patial, Pankaj Markand Kulurkar, Avnesh Kumari, Mahesh S, Yogendra S Padwad, Sudesh Kumar Yadav
ABSTRACT
The ever increasing incidences of non-healing skin wounds have paved way for many efforts on the convoluted process of wound healing. Unfortunately, the lack of relevance and success of modern wound dressings in healing of acute and diabetic wounds still remains a matter of huge concern. Here, an in situ three step approach was embraced for the development of nanocomposite (NCs) dressings by impregnating silver nanoparticles (AgNPs) onto a matrix of cellulose nanocrystals (CNCs) isolated from Syzygium cumini leaves using an environmental friendly approach. Topical application of NCs (ointments and strips) on acute and diabetic wounds of mice documented enhanced tissue repair (~99% wound closure) via decrease in inflammation; increase in angiogenesis, collagen deposition, and rate of neo-epithelialization that ultimately led to formation of aesthetically sound skin in lesser time than controls. Due to the synergistic action of CNCs (having high water uptake capacity) and AgNPs (anti-microbial agents), NCs tend to increase the expression of essential growth factors (FGF, PDGF and VEGF) and collagen while decreasing the pro-inflammatory factors (IL-6 and TNF-α) at the same time, thus accelerating healing. The results suggested the potential of these developed anti-microbial, cytocompatible and nanoporous NCs having optimized AgNPs concentration as ideal dressings for effective wound management.