Merck
Autophagy-Sirt3 axis decelerates hematopoietic aging.
Aging cell (2020-09-21)
Yixuan Fang, Ni An, Lingjiang Zhu, Yue Gu, Jiawei Qian, Gaoyue Jiang, Ruijin Zhao, Wen Wei, Li Xu, Gaochuan Zhang, Xingyun Yao, Na Yuan, Suping Zhang, Yun Zhao, Jianrong Wang
ABSTRACT
Autophagy suppresses mitochondrial metabolism to preserve hematopoietic stem cells (HSCs) in mice. However, the mechanism by which autophagy regulates hematopoietic aging, in particular in humans, has largely been unexplored. Here, we demonstrate that reduction of autophagy in both hematopoietic cells and their stem cells is associated with aged hematopoiesis in human population. Mechanistically, autophagy delays hematopoietic aging by activating the downstream expression of Sirt3, a key mitochondrial protein capable of rejuvenating blood. Sirt3 is the most abundant Sirtuin family member in HSC-enriched population, though it declines as the capacity for autophagy deteriorates with aging. Activation of autophagy upregulates Sirt3 in wild-type mice, whereas in autophagy-defective mice, Sirt3 expression is crippled in the entire hematopoietic hierarchy, but forced expression of Sirt3 in HSC-enriched cells reduces oxidative stress and prevents accelerated hematopoietic aging from autophagy defect. Importantly, the upregulation of Sirt3 by manipulation of autophagy is validated in human HSC-enriched cells. Thus, our results identify an autophagy-Sirt3 axis in regulating hematopoietic aging and suggest a possible interventional solution to human blood rejuvenation via activation of the axis.
Sigma-Aldrich
Saponin from quillaja bark, Sapogenin content ≥10 %
Sigma-Aldrich
Bafilomycin A1, Streptomyces griseus, Bafilomycin A1, CAS 88899-55-2, acts as a highly potent and specific inhibitor of vacuolar-type H+-ATPase (Ki = 500 pM). Blocks the fusion of autophagosome with lysosome.
Sigma-Aldrich
BrdU (5-Bromo-2′-deoxyuridine), Used for the study of DNA synthesis where it is incorporated into DNA in place of thymidine, BrdU can be used in conjunction with anti-BrdU for immunocytochemical analysis of cell proliferation.