Merck
Bisindolylmaleimides in anti-cancer therapy - more than PKC inhibitors.
Advances in medical sciences (2008-07-19)
B Pajak, S Orzechowska, B Gajkowska, A Orzechowski
ABSTRACT
Bisindolylmaleimide derivatives were originally described as protein kinase C inhibitors. However, several studies have shown that bisindolylmaleimides target several other signaling molecules. The review presents bisindolylmaleimide-mediated PKC-dependent and PKC-independent biological effects, such as reversal of MDR and modulation of Wnt signaling through GSK-3b and b-catenin. Importantly, the potent proapoptotic properties of bisindolylmaleimides are also described. Bis-IX appears as the most efficient activator of intrinsic apoptotic pathway and additionally, facilitates extrinsic apoptosis. Presented molecular mechanisms indicate that bisindolylmaleimides could be useful agents in anticancer therapy. They repress uncontrolled proliferation and restore the sensitivity to chemotherapy which allows eradication of cancer cells.
Sigma-Aldrich
Bisindolylmaleimide IV, ≥98% (TLC), solid