Merck
  • Functional expression of formyl peptide receptor family in human NK cells.
Functional expression of formyl peptide receptor family in human NK cells.
Journal of immunology (Baltimore, Md. : 1950) (2009-10-22)
Sang Doo Kim, Jung Mo Kim, Seong Ho Jo, Ha Young Lee, Sun Young Lee, Jae Woong Shim, Su-Kil Seo, Jeanho Yun, Yoe-Sik Bae
ABSTRACT
We determined the expression of the formyl peptide receptor (FPR) family and the functional roles of the FPR family in NK cells. All tested human NK cells express two members of the FPR family (FPR1 and FPR2). The expression of FPR3 was noted to occur in a donor-specific manner. The stimulation of NK cells with FPR family-selective agonists (fMLF (N-formyl-Met-Leu-Phe), MMK-1, F2L, and WKYMVm (Trp-Lys-Tyr-Met-Val-d-Met)) elicited cytolytic activity in resting NK cells, but not in IL-2-activated NK cells; the cytolytic activity was not inhibited by pertussis toxin. The FPR family agonists also stimulated chemotactic migration of IL-2-activated NK cells, but not resting NK cells; the chemotactic migration was completely inhibited by pertussis toxin. WKYMVm stimulates ERK, p38 MAPK, and JNK activities in both resting and IL-2-activated NK cells. WKYMVm-induced chemotactic migration was partially inhibited by PD98059 (2'-amino-3'-methoxyflavone); however, the inhibition of JNK by its selective inhibitor (SP600125, anthra[1,9-cd]pyrazol-6(2H)-one) dramatically inhibited the WKYMVm-induced cytolytic activity. Furthermore, WKYMVm-induced chemotactic migration and cytolytic activity were partly inhibited by FPR family-selective antagonists (cyclosporin H and WRWWWW). Taken together, our findings indicate that human NK cells express functional members of the FPR family, and in turn the activation of the three members of the FPR receptor family elicit cytolytic activity in NK cells, thus suggesting that the receptors are potentially important therapeutic targets for the modulation of NK cell-mediated immune responses.
Sigma-Aldrich
MMK-1 trifluoroacetate, ≥95% (HPLC)