Merck
  • Effect of different exogenous fatty acids on the cytosolic triacylglycerol content in bovine mammary cells.
Effect of different exogenous fatty acids on the cytosolic triacylglycerol content in bovine mammary cells.
Animal nutrition (Zhongguo xu mu shou yi xue hui) (2019-06-14)
Einar Vargas-Bello-Pérez, Juan J Loor, Philip C Garnsworthy
ABSTRACT
The objective of this study was to determine how cytosolic triacylglycerols (TAG) are stored in mammary cells and whether this depends on the individual chemical configuration of fatty acids (FA). This objective was accomplished by addition of different FA to a FA-free medium used to culture mammary alveolar cells-large T antigen cells (MAC-T). Treatments consisted of adding FA (palmitate, stearate, oleate, linoleate, rumenic acid [CLA], elaidate and vaccinate) solutions to the medium at 100, 200, 300 and 400 mmol/L concentrations for a 24-h incubation period. At the end of each incubation period, cytosolic TAG, DNA and protein contents were measured. Palmitate, vaccenate, linoleate and CLA increased (P < 0.05) cytosolic TAG (μg/mg protein). Palmitate and CLA increased (P < 0.05) cytosolic TAG adjusted for DNA content. Overall, effects on cytosolic TAG accumulation depended on individual FA structure (chain length, degree of saturation, and number and orientation of FA double bonds). In addition, the long-chain FA used in this study did not have a detrimental effect on MAC-T cells as indicated by cytosolic protein and DNA contents reflecting their biological role in lipid accumulation.
Sigma-Aldrich
apo-Transferrin bovine, BioReagent, suitable for cell culture, ≥98%