Merck
  • Carbonic anhydrase activators: Activation of the human cytosolic isozyme III and membrane-associated isoform IV with amino acids and amines.
Carbonic anhydrase activators: Activation of the human cytosolic isozyme III and membrane-associated isoform IV with amino acids and amines.
Bioorganic & medicinal chemistry letters (2008-07-17)
Daniela Vullo, Isao Nishimori, Andrea Scozzafava, Claudiu T Supuran
ABSTRACT
An activation study of the human carbonic anhydrase (hCA, EC 4.2.1.1) isoforms hCA III (cytosolic) and IV (membrane-associated) with a series of natural and non-natural amino acids and aromatic/heterocyclic amines is reported. hCA III was efficiently activated by d-His, serotonin, pyridyl-alkylamines, and aminoethyl-piperazine/morpholine (K(A)s of 91nM-1.12microM), whereas the best hCA IV activators were 4-amino-phenylalanine, serotonin, and 4-(2-aminoethyl)-morpholine (K(A)s of 79nM-3.14microM). Precise steric and electronic requirements are needed to be present in the molecules of effective CA III/IV activators, in order to assure an adequate fit within the enzyme active site for the formation of the enzyme-activator complex, and for efficient proton transfer processes between the active site and the reaction medium. The activation profiles of CA III and IV are distinct from those of all other mammalian CA isoforms investigated so far for their interaction with amino acids and amines.
Product Description
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Histidine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
1-(2-Aminoethyl)piperazine, 99%
Sigma-Aldrich
D-Tryptophan, ≥98.0% (HPLC)
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Histamine, ≥97.0%
Sigma-Aldrich
D-Histidine, ≥98% (TLC)
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
D-Phenylalanine, ≥98% (HPLC)
SAFC
L-Histidine
SAFC
L-Tyrosine
Sigma-Aldrich
2-(2-Pyridyl)ethylamine, 95%
Sigma-Aldrich
2-Picolylamine, 99%
Sigma-Aldrich
L-Phenylalanine, 99%, FCC
Sigma-Aldrich
4-(2-Aminoethyl)morpholine, 99%
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Phenylalanine, reagent grade, ≥98%
Sigma-Aldrich
3,4-Dihydroxy-D-phenylalanine, powder, ≥95%
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)
Supelco
L-Tryptophan, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Phenylalanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Phenylalanine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Phenylalanine, BioUltra, ≥99.0% (NT)