Merck
  • CLP290 promotes the sedative effects of midazolam in neonatal rats in a KCC2-dependent manner: A laboratory study in rats.
CLP290 promotes the sedative effects of midazolam in neonatal rats in a KCC2-dependent manner: A laboratory study in rats.
PloS one (2021-03-13)
Akiko Doi, Tomoyuki Miyazaki, Takahiro Mihara, Maiko Ikeda, Ryo Niikura, Tomio Andoh, Takahisa Goto
ABSTRACT
Immature neurons dominantly express the Na+-K+-2Cl- cotransporter isoform 1 (NKCC1) rather than the K+-Cl- cotransporter isoform 2 (KCC2). The intracellular chloride ion concentration ([Cl-]i) is higher in immature neurons than in mature neurons; therefore, γ-aminobutyric acid type A (GABAA) receptor activation in immature neurons does not cause chloride ion influx and subsequent hyperpolarization. In our previous work, we found that midazolam, benzodiazepine receptor agonist, causes less sedation in neonatal rats compared to adult rats and that NKCC1 blockade by bumetanide enhances the midazolam-induced sedation in neonatal, but not in adult, rats. These results suggest that GABA receptor activation requires the predominance of KCC2 over NKCC1 to exert sedative effects. In this study, we focused on CLP290, a novel KCC2-selective activator, and found that midazolam administration at 20 mg/kg after oral CLP290 intake significantly prolonged the righting reflex latency even in neonatal rats at postnatal day 7. By contrast, CLP290 alone did not exert sedative effects. Immunohistochemistry showed that midazolam combined with CLP290 decreased the number of phosphorylated cAMP response element-binding protein-positive cells in the cerebral cortex, suggesting that CLP290 reverted the inhibitory effect of midazolam. Moreover, the sedative effect of combined CLP290 and midazolam treatment was inhibited by the administration of the KCC2-selective inhibitor VU0463271, suggesting indirectly that the sedation-promoting effect of CLP290 was mediated by KCC2 activation. To our knowledge, this study is the first report showing the sedation-promoting effect of CLP290 in neonates and providing behavioral and histological evidence that CLP290 reverted the sedative effect of GABAergic drugs through the activation of KCC2. Our data suggest that the clinical application of CLP290 may provide a breakthrough in terms of midazolam-resistant sedation.
Sigma-Aldrich
Anti-phospho-CREB (Ser133) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-K+/Cl- Cotransporter (KCC2) Antibody, Upstate®, from rabbit
Sigma-Aldrich
CLP290, ≥98% (HPLC)