Merck
  • Inhibitor scaffolds for 2-oxoglutarate-dependent histone lysine demethylases.
Inhibitor scaffolds for 2-oxoglutarate-dependent histone lysine demethylases.
Journal of medicinal chemistry (2008-10-24)
Nathan R Rose, Stanley S Ng, Jasmin Mecinović, Benoît M R Liénard, Simon H Bello, Zhe Sun, Michael A McDonough, Udo Oppermann, Christopher J Schofield
ABSTRACT
The dynamic methylation of histone lysyl residues plays an important role in biology by regulating transcription, maintaining genomic integrity, and by contributing to epigenetic effects. Here we describe a variety of inhibitor scaffolds that inhibit the human 2-oxoglutarate-dependent JMJD2 subfamily of histone demethylases. Combined with structural data, these chemical starting points will be useful to generate small-molecule probes to analyze the physiological roles of these enzymes in epigenetic signaling.
Sigma-Aldrich
2,6-Pyridinedicarboxylic acid, 99%
Supelco
2,6-Pyridinedicarboxylic acid, suitable for ion chromatography, ≥99.5% (T)
Sigma-Aldrich
2,3-Pyridinedicarboxylic acid, 99%
Sigma-Aldrich
Succinic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Butyric acid, ≥99%
Sigma-Aldrich
Butyric acid, ≥99%, FG
Sigma-Aldrich
Fumaric acid, ≥99.0% (T)
Sigma-Aldrich
Succinic acid, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Succinic acid, puriss. p.a., ACS reagent, ≥99.5% (T)
Sigma-Aldrich
Fumaric acid, FCC, FG
Sigma-Aldrich
2,5-Pyridinedicarboxylic acid, 98%
Sigma-Aldrich
Butyric acid, natural, ≥99%, FCC, FG
Sigma-Aldrich
2,2′-Bipyridine-4,4′-dicarboxylic acid, 98%
Supelco
Butyric acid, analytical standard
Sigma-Aldrich
Succinic acid, ReagentPlus®, BioRenewable, ≥99.0%
Sigma-Aldrich
Succinic acid, BioReagent, BioRenewable, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Succinic acid, BioXtra, BioRenewable, ≥99.0%