Merck
  • Spatial alterations of De Novo purine biosynthetic enzymes by Akt-independent PDK1 signaling pathways.
Spatial alterations of De Novo purine biosynthetic enzymes by Akt-independent PDK1 signaling pathways.
PloS one (2018-04-19)
Danielle L Schmitt, Anand Sundaram, Miji Jeon, Bao Tran Luu, Songon An
ABSTRACT
A macromolecular complex of the enzymes involved in human de novo purine biosynthesis, the purinosome, has been shown to consist of a core assembly to regulate the metabolic activity of the pathway. However, it remains elusive whether the core assembly itself can be selectively controlled in the cytoplasm without promoting the purinosome. Here, we reveal that pharmacological inhibition of the cytoplasmic activity of 3-phosphoinositide-dependent protein kinase 1 (PDK1) selectively promotes the formation of the core assembly, but not the purinosome, in cancer cells. However, alternative signaling cascades that are associated with the plasma membrane-bound PDK1 activity, including Akt-mediated cascades, regulate neither the core assembly nor the purinosome in our conditions. Along with immunofluorescence microscopy and a knock-down study against PDK1 using small interfering RNAs, we reveal that cytoplasmic PDK1-associated signaling pathways regulate subcellular colocalization of three enzymes that form the core assembly of the purinosome in an Akt-independent manner. Collectively, this study reveals a new mode of compartmentalization of purine biosynthetic enzymes controlled by spatially resolved signaling pathways.
Sigma-Aldrich
GSK2334470, ≥98% (HPLC)
Sigma-Aldrich
Akt Inhibitor VI, Akt-in, The Akt Inhibitor VI, Akt-in controls the biological activity of Akt. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
LY 294002, LY294002, CAS 154447-36-6, is a cell-permeable, potent, reversible, and specific inhibitor of PI 3-kinase ((IC₅₀ = 1.4 µM). Acts on the ATP-binding site.
Sigma-Aldrich
Rapamycin, ≥95% (HPLC), powder
Sigma-Aldrich
Akt Inhibitor X, The Akt Inhibitor X, also referenced under CAS 925681-41-0, controls the biological activity of Akt. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.