Merck
  • mTORC1 directly inhibits AMPK to promote cell proliferation under nutrient stress.
mTORC1 directly inhibits AMPK to promote cell proliferation under nutrient stress.
Nature metabolism (2020-01-30)
Naomi X Y Ling, Adrian Kaczmarek, Ashfaqul Hoque, Elizabeth Davie, Kevin R W Ngoei, Kaitlin R Morrison, William J Smiles, Gabriella M Forte, Tingting Wang, Shervi Lie, Toby A Dite, Christopher G Langendorf, John W Scott, Jonathan S Oakhill, Janni Petersen
ABSTRACT
Central to cellular metabolism and cell proliferation are highly conserved signalling pathways controlled by mammalian target of rapamycin (mTOR) and AMP-activated protein kinase (AMPK)1,2, dysregulation of which are implicated in pathogenesis of major human diseases such as cancer and type 2 diabetes. AMPK pathways leading to reduced cell proliferation are well established and, in part, act through inhibition of TOR complex-1 (TORC1) activity. Here we demonstrate reciprocal regulation, specifically that TORC1 directly down-regulates AMPK signalling by phosphorylating the evolutionarily conserved residue Ser367 in the fission yeast AMPK catalytic subunit Ssp2, and AMPK α1Ser347/α2Ser345 in the mammalian homologs, which is associated with reduced phosphorylation of activation loop Thr172. Genetic or pharmacological inhibition of TORC1 signalling led to AMPK activation in the absence of increased AMP:ATP ratios; under nutrient stress conditions this was associated with growth limitation in both yeast and human cell cultures. Our findings reveal fundamental, bi-directional regulation between two major metabolic signalling networks and uncover new opportunity for cancer treatment strategies aimed at suppressing cell proliferation in the nutrient-poor tumor microenvironment.
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), crystalline
Sigma-Aldrich
LKB1/MO25/STRAD Active human, recombinant, expressed in baculovirus infected insect cells, ≥70% (SDS-PAGE)
Millipore
EZview Red Anti-HA Affinity Gel
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
SBI-0206965, ≥98% (HPLC)
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)